missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Lenalidomide 4'-PEG2-azide
Description
Lenalidomide 4'-PEG2-azide is a functionalized cereblon ligand for PROTAC® research and development; incorporates an E3 ligase ligand plus a PEG2 linker with terminal azide ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | 3-[4-[[2-[2-(2-Azidoethoxy)ethoxy]ethyl]amino]-1,3-dihydro-1-oxo-2 H-isoindol-2-yl]-2,6-piperidinedione |
| CAS | 2399455-48-0 |
| Quantity | 25 mg |
| Product Line | Tocris |
| Target | Degrader Building Blocks |
| Molecular Formula | C19H24N6O5 |
| Purity | 0.95 |