missing translation for 'onlineSavingsMsg'
Learn More

Leflunomide

Catalog No. AAJ65917MC
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ65917MC 100 mg
AAJ65917ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ65917MC Supplier Thermo Scientific Chemicals Supplier No. J65917MC

CAS: 75706-12-6 | C12H9F3N2O2 | 270.211 g/mol

An immunosuppressive agent. Inhibits T and B cell proliferation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • An immunosuppressive agent
  • Inhibits T and B cell proliferation

Chemical Identifiers

CAS 75706-12-6
Molecular Formula C12H9F3N2O2
Molecular Weight (g/mol) 270.211
MDL Number MFCD00867593
InChI Key VHOGYURTWQBHIL-UHFFFAOYSA-N
Synonym leflunomide, arava, leflunomidum, lefunamide, leflunomida, leflunomidum inn-latin, 5-methyl-n-4-trifluoromethyl phenyl-1,2-oxazole-4-carboxamide, leflunomida inn-spanish, su 101 pharmaceutical, arava tn
PubChem CID 3899
ChEBI CHEBI:6402
IUPAC Name 5-methyl-N-[4-(trifluoromethyl)phenyl]-1,2-oxazole-4-carboxamide
SMILES CC1=C(C=NO1)C(=O)NC2=CC=C(C=C2)C(F)(F)F

Specifications

Color White
Quantity 100 mg
UN Number UN2811
Merck Index 14,5432
Formula Weight 270.21
Physical Form Crystalline Solid
Chemical Name or Material Leflunomide
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECSNumber NY2354200
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only