missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Leflunomide | |
| 168°C | |
| C12H9F3N2O2 | |
| 1 g | |
| leflunomide, arava, leflunomidum, lefunamide, leflunomida, leflunomidum inn-latin, 5-methyl-n-4-trifluoromethyl phenyl-1,2-oxazole-4-carboxamide, leflunomida inn-spanish, su 101 pharmaceutical, arava tn | |
| CC1=C(C=NO1)C(=O)NC2=CC=C(C=C2)C(F)(F)F | |
| 270.211 | |
| CHEBI:6402 | |
| ≥98.0% (HPLC,N) |
| 75706-12-6 | |
| White | |
| MFCD00867593 | |
| 2811 | |
| VHOGYURTWQBHIL-UHFFFAOYSA-N | |
| 5-methyl-N-[4-(trifluoromethyl)phenyl]-1,2-oxazole-4-carboxamide | |
| 3899 | |
| 270.21 | |
| Crystalline Powder |
Chemical Identifiers
| 75706-12-6 | |
| 270.211 | |
| VHOGYURTWQBHIL-UHFFFAOYSA-N | |
| 3899 | |
| 5-methyl-N-[4-(trifluoromethyl)phenyl]-1,2-oxazole-4-carboxamide |
| C12H9F3N2O2 | |
| MFCD00867593 | |
| leflunomide, arava, leflunomidum, lefunamide, leflunomida, leflunomidum inn-latin, 5-methyl-n-4-trifluoromethyl phenyl-1,2-oxazole-4-carboxamide, leflunomida inn-spanish, su 101 pharmaceutical, arava tn | |
| CHEBI:6402 | |
| CC1=C(C=NO1)C(=O)NC2=CC=C(C=C2)C(F)(F)F |
Safety and Handling
RTECSNumber : NY2354200
TSCA : No
Recommended Storage : Refrigerator