missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LEF1 Monoclonal Antibody (REMB6)
Mouse Monoclonal Antibody
$823.92 - $1342.39
Specifications
| Antigen | LEF1 |
|---|---|
| Clone | REMB6 |
| Concentration | 1 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
01-674-297
|
Invitrogen™
MA112420 |
250 μg |
Each for $1,342.39
|
|
|||||
|
01-674-298
|
Invitrogen™
MA112421 |
100 μg |
Each for $823.92
|
|
|||||
Description
A suggested positive control for this product is Jurkat cell lysate.
This gene encodes a transcription factor belonging to a family of proteins that share homology with the high mobility group protein-1. The protein encoded by this gene can bind to a functionally important site in the T-cell receptor-alpha enhancer, thereby conferring maximal enhancer activity. This transcription factor is involved in the Wnt signaling pathway, and it may function in hair cell differentiation and follicle morphogenesis. Mutations in this gene have been found in somatic sebaceous tumors. This gene has also been linked to other cancers, including androgen-independent prostate cancer. Alternative splicing results in multiple transcript variants.Specifications
| LEF1 | |
| 1 mg/mL | |
| ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 0.08% sodium azide | |
| P27782, Q9UJU2 | |
| 16842, 51176 | |
| Full-length recombinant LEF-1 protein. | |
| Antibody |
| REMB6 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Monoclonal | |
| Liquid | |
| IgG1 | |
| Human, Mouse | |
| LEF1 | |
| 3000002B05; AI451430; DKFZp586H0919; FLJ46390; HGNC:6551; HMG Binding Domain; LEF1; Lef-1; lymphoid enhancer binding factor 1; lymphoid enhancer binding factor 1 short isoform; lymphoid enhancer-binding factor 1; OTTHUMP00000219736; OTTHUMP00000219737; T cell-specific transcription factor 1-alpha; TCF10; TCF1ALPHA; TCF1-alpha; TCF7L3 | |
| LEF1 | |
| Primary | |
| Ammonium sulfate precipitation |