missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LEF1 Monoclonal Antibody (REMB6)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA112420
Description
A suggested positive control for this product is Jurkat cell lysate.
This gene encodes a transcription factor belonging to a family of proteins that share homology with the high mobility group protein-1. The protein encoded by this gene can bind to a functionally important site in the T-cell receptor-alpha enhancer, thereby conferring maximal enhancer activity. This transcription factor is involved in the Wnt signaling pathway, and it may function in hair cell differentiation and follicle morphogenesis. Mutations in this gene have been found in somatic sebaceous tumors. This gene has also been linked to other cancers, including androgen-independent prostate cancer. Alternative splicing results in multiple transcript variants.Specifications
| LEF1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.08% sodium azide | |
| P27782, Q9UJU2 | |
| LEF1 | |
| Full-length recombinant LEF-1 protein. | |
| 250 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| REMB6 | |
| Unconjugated | |
| LEF1 | |
| 3000002B05; AI451430; DKFZp586H0919; FLJ46390; HGNC:6551; HMG Binding Domain; LEF1; Lef-1; lymphoid enhancer binding factor 1; lymphoid enhancer binding factor 1 short isoform; lymphoid enhancer-binding factor 1; OTTHUMP00000219736; OTTHUMP00000219737; T cell-specific transcription factor 1-alpha; TCF10; TCF1ALPHA; TCF1-alpha; TCF7L3 | |
| Mouse | |
| Ammonium sulfate precipitation | |
| RUO | |
| 16842, 51176 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |