missing translation for 'onlineSavingsMsg'
Learn More

Lead Nitrate, Crystal, ACS, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor L107550KGBL

 View more versions of this product

Catalog No. 18611239

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

10099-74-8
50 kg
ACS
1
99%
Fiber Drum

Chemical Identifiers

10099-74-8
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.