missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lawesson's Reagent 95.0+%, TCI America™
$75.95 - $459.46
Chemical Identifiers
| CAS | 19172-47-5 |
|---|---|
| Molecular Formula | C14H14O2P2S4 |
| Molecular Weight (g/mol) | 404.452 |
| MDL Number | MFCD00005171 |
| InChI Key | CFHGBZLNZZVTAY-UHFFFAOYSA-N |
| Synonym | lawesson's reagent, lawesson reagent, 2,4-bis 4-methoxyphenyl-1,3,2,4-dithiadiphosphetane 2,4-disulfide, unii-a4125mq8rx, 1,3,2,4-dithiadiphosphetane, 2,4-bis 4-methoxyphenyl-, 2,4-disulfide, 2,4-bis 4-methoxyphenyl-2,4-dithioxo-1,3,2,4-dithiadiphosphetane, 2,4-bis 4-methoxyphenyl-1,3,2,4-dithiadiphosphetane-2,4-disulfide, 2,4-bis 4-methoxyphenyl-1,3-dithia-2,4-diphosphetane 2,4-disulfide, 2,4-bis 4-methoxyphenyl-1,3-dithia-2,4-diphosphetane 2,4-disulphide, 4-methoxyphenylthiophosphoric cyclic di thioanhydride |
| PubChem CID | 87949 |
| IUPAC Name | 2,4-bis(4-methoxyphenyl)-2,4-bis(sulfanylidene)-1,3,2$l^{5},4$l^{5}-dithiadiphosphetane |
| SMILES | COC1=CC=C(C=C1)P2(=S)SP(=S)(S2)C3=CC=C(C=C3)OC |
Chemical Identifiers
| 19172-47-5 | |
| 404.452 | |
| CFHGBZLNZZVTAY-UHFFFAOYSA-N | |
| 87949 | |
| COC1=CC=C(C=C1)P2(=S)SP(=S)(S2)C3=CC=C(C=C3)OC |
| C14H14O2P2S4 | |
| MFCD00005171 | |
| lawesson's reagent, lawesson reagent, 2,4-bis 4-methoxyphenyl-1,3,2,4-dithiadiphosphetane 2,4-disulfide, unii-a4125mq8rx, 1,3,2,4-dithiadiphosphetane, 2,4-bis 4-methoxyphenyl-, 2,4-disulfide, 2,4-bis 4-methoxyphenyl-2,4-dithioxo-1,3,2,4-dithiadiphosphetane, 2,4-bis 4-methoxyphenyl-1,3,2,4-dithiadiphosphetane-2,4-disulfide, 2,4-bis 4-methoxyphenyl-1,3-dithia-2,4-diphosphetane 2,4-disulfide, 2,4-bis 4-methoxyphenyl-1,3-dithia-2,4-diphosphetane 2,4-disulphide, 4-methoxyphenylthiophosphoric cyclic di thioanhydride | |
| 2,4-bis(4-methoxyphenyl)-2,4-bis(sulfanylidene)-1,3,2$l^{5},4$l^{5}-dithiadiphosphetane |
Specifications
| Undesignated | |
| 230°C | |
| MFCD00005171 | |
| CFHGBZLNZZVTAY-UHFFFAOYSA-N | |
| 2,4-bis(4-methoxyphenyl)-2,4-bis(sulfanylidene)-1,3,2$l^{5},4$l^{5}-dithiadiphosphetane | |
| 87949 | |
| ≥95.0% (T) | |
| Crystal-Powder at 20°C |
| 19172-47-5 | |
| C14H14O2P2S4 | |
| lawesson's reagent, lawesson reagent, 2,4-bis 4-methoxyphenyl-1,3,2,4-dithiadiphosphetane 2,4-disulfide, unii-a4125mq8rx, 1,3,2,4-dithiadiphosphetane, 2,4-bis 4-methoxyphenyl-, 2,4-disulfide, 2,4-bis 4-methoxyphenyl-2,4-dithioxo-1,3,2,4-dithiadiphosphetane, 2,4-bis 4-methoxyphenyl-1,3,2,4-dithiadiphosphetane-2,4-disulfide, 2,4-bis 4-methoxyphenyl-1,3-dithia-2,4-diphosphetane 2,4-disulfide, 2,4-bis 4-methoxyphenyl-1,3-dithia-2,4-diphosphetane 2,4-disulphide, 4-methoxyphenylthiophosphoric cyclic di thioanhydride | |
| COC1=CC=C(C=C1)P2(=S)SP(=S)(S2)C3=CC=C(C=C3)OC | |
| 404.452 | |
| 404.45 | |
| 25 g | |
| Lawesson′s Reagent [Sulfurating Reagent] |
Safety and Handling
TSCA : Yes