accessibility menu, dialog, popup

UserName

CAS RN 19172-47-5

IUPAC Name: bis(4-methoxyphenyl)-1,3,2λ⁵,4λ⁵-dithiadiphosphetane-2,4-dithione
Synonyms: lawesson's reagent
Molecular Weight (g/mol): 404.45
Molecular Formula: C14H14O2P2S4
InChi Key: CFHGBZLNZZVTAY-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)P1(=S)SP(=S)(S1)C1=CC=C(OC)C=C1
Percent Purity 
  • (2)
  • (4)
  • (3)
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (7)
  • (2)
  • (4)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)