missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lauramide 96.0+%, TCI America™
$152.86 - $1493.11
Chemical Identifiers
| CAS | 1120-16-7 |
|---|---|
| Molecular Formula | C12H25NO |
| Molecular Weight (g/mol) | 199.338 |
| MDL Number | MFCD00025532 |
| InChI Key | ILRSCQWREDREME-UHFFFAOYSA-N |
| Synonym | Dodecanamide |
| PubChem CID | 14256 |
| ChEBI | CHEBI:34726 |
| IUPAC Name | dodecanamide |
| SMILES | CCCCCCCCCCCC(=O)N |
Chemical Identifiers
| 1120-16-7 | |
| 199.338 | |
| ILRSCQWREDREME-UHFFFAOYSA-N | |
| 14256 | |
| dodecanamide |
| C12H25NO | |
| MFCD00025532 | |
| Dodecanamide | |
| CHEBI:34726 | |
| CCCCCCCCCCCC(=O)N |
Specifications
| 1120-16-7 | |
| White-Yellow | |
| C12H25NO | |
| 25 g | |
| ILRSCQWREDREME-UHFFFAOYSA-N | |
| dodecanamide | |
| 14256 | |
| 199.34 | |
| Crystalline Powder |
| 99°C | |
| 200°C | |
| MFCD00025532 | |
| Dodecanamide | |
| CCCCCCCCCCCC(=O)N | |
| 199.338 | |
| CHEBI:34726 | |
| ≥96.0% (GC) | |
| Lauramide |
Safety and Handling
EINECSNumber : (2)-0824
TSCA : Yes