missing translation for 'onlineSavingsMsg'
Learn More

Lauramide 96.0+%, TCI America™

Supplier:  TCI America L0077100G

 View more versions of this product

Catalog No. L0077100G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Lauramide
99°C
200°C
MFCD00025532
Dodecanamide
CCCCCCCCCCCC(=O)N
199.338
CHEBI:34726
≥96.0% (GC)
1120-16-7
White-Yellow
C12H25NO
100 g
ILRSCQWREDREME-UHFFFAOYSA-N
dodecanamide
14256
199.34
Crystalline Powder

Chemical Identifiers

1120-16-7
199.338
ILRSCQWREDREME-UHFFFAOYSA-N
14256
dodecanamide
C12H25NO
MFCD00025532
Dodecanamide
CHEBI:34726
CCCCCCCCCCCC(=O)N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0824

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions