missing translation for 'onlineSavingsMsg'
Learn More

PIG™ Latching Drum Lid with Fast-Latch Ring

Catalog No. 18366499
Change view
Click to view available options
Color:
Black
Yellow
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
18366499 Yellow
18366497 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18366499 Supplier PIG™ Supplier No. DRM1072YW

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Latching drum lid to keep drums and hazardous waste storage containers securely closed and locked.

  • For 55 gal. Steel Drums
  • Fast Latch
  • Fast-latch ring for easy installation
  • Adjustment rod to expand ring to fit reconditioned drums with larger diameters
  • Latching handle easily opens and closes with one hand
  • Nitrile gasket and locking mechanism seal lid to help reduce vapor emissions and keep drum contents dry and secure
  • Easy-access lid closes securely to help you meet hazardous waste regulations
  • Hinge stops keep funnel lid upright when open
  • Durable, chemical-resistant powder coat helps ensure long service life
  • Lockable latch helps prevent unauthorized access
  • Hinge pin is made for easy lid removal without loosening band around drum
  • Multiple lid color options let you segregate drums for safe, compliant filling

Specifications

Product Type Latching Drum Lid
Color Yellow
Sterility Non-sterile
Length (English) 26.75 in.
Length (Metric) 68 cm
Height (English) 4.37 in.
Height (Metric) 11 cm
Diameter (English) Inner 23.25 in.
Diameter (Metric) Inner 59 cm