missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lappaconitine Hydrobromide 95.0+%, TCI America™
Supplier: TCI America L02721G
Specifications
| Lappaconitine Hydrobromide | |
| 222°C | |
| C32H45BrN2O8 | |
| 1 g | |
| DUHCHCGKLJHQCG-JOJFFJMOSA-N | |
| (2S,3S,5S,8S,10R,17R)-13-[2-(2-acetamidophenyl)-2-oxoethyl]-3,5,8-trihydroxy-4,6,16-trimethoxy-11-methyl-11-azahexacyclo[7.7.2.1²,âµ.0¹,¹â°.0³,â¸.0¹³,¹â·]nonadecan-11-ium bromide | |
| 134129700 | |
| ≥95.0% (N) |
| 97792-45-5 | |
| White-Yellow | |
| MFCD00171334 | |
| 1544 | |
| [Br-].COC1[C@]2(O)C[C@@H]3[C@@]1(O)[C@](O)(CC2OC)C1C[C@H]2C33[C@@H]1[NH+](C)CC2(CC(=O)C1=CC=CC=C1NC(C)=O)CCC3OC | |
| 665.62 | |
| 665.62 | |
| Crystalline Powder |
Chemical Identifiers
| 97792-45-5 | |
| 665.62 | |
| DUHCHCGKLJHQCG-JOJFFJMOSA-N | |
| (2S,3S,5S,8S,10R,17R)-13-[2-(2-acetamidophenyl)-2-oxoethyl]-3,5,8-trihydroxy-4,6,16-trimethoxy-11-methyl-11-azahexacyclo[7.7.2.1²,âµ.0¹,¹â°.0³,â¸.0¹³,¹â·]nonadecan-11-ium bromide |
| C32H45BrN2O8 | |
| MFCD00171334 | |
| 134129700 | |
| [Br-].COC1[C@]2(O)C[C@@H]3[C@@]1(O)[C@](O)(CC2OC)C1C[C@H]2C33[C@@H]1[NH+](C)CC2(CC(=O)C1=CC=CC=C1NC(C)=O)CCC3OC |
Safety and Handling
RTECSNumber : AR5569600
TSCA : No
Recommended Storage : Refrigerator