missing translation for 'onlineSavingsMsg'
Learn More

Lappaconitine Hydrobromide 95.0+%, TCI America™

Catalog No. p-7131903
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L0272200MG 200 mg
L02721G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L0272200MG Supplier TCI America Supplier No. L0272200MG

Chemical Identifiers

CAS 97792-45-5
Molecular Formula C32H45BrN2O8
Molecular Weight (g/mol) 665.62
MDL Number MFCD00171334
InChI Key DUHCHCGKLJHQCG-JOJFFJMOSA-N
PubChem CID 134129700
IUPAC Name (2S,3S,5S,8S,10R,17R)-13-[2-(2-acetamidophenyl)-2-oxoethyl]-3,5,8-trihydroxy-4,6,16-trimethoxy-11-methyl-11-azahexacyclo[7.7.2.1²,⁵.0¹,¹⁰.0³,⁸.0¹³,¹⁷]nonadecan-11-ium bromide
SMILES [Br-].COC1[C@]2(O)C[C@@H]3[C@@]1(O)[C@](O)(CC2OC)C1C[C@H]2C33[C@@H]1[NH+](C)CC2(CC(=O)C1=CC=CC=C1NC(C)=O)CCC3OC

Specifications

Melting Point 222°C
Color White-Yellow
Quantity 200 mg
UN Number 1544
Formula Weight 665.62
Percent Purity ≥95.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Lappaconitine Hydrobromide
RTECSNumber AR5569600
TSCA No
Recommended Storage Refrigerator