accessibility menu, dialog, popup

UserName

CAS RN 97792-45-5

BrH3CONHCH3(R)(R)(S)(S)OH(S)OH(S)OHOCH3OCH3ONHH3CO

CAS RN 97792-45-5

IUPAC Name: (2S,3S,5S,8S,10R,17R)-13-[2-(2-acetamidophenyl)-2-oxoethyl]-3,5,8-trihydroxy-4,6,16-trimethoxy-11-methyl-11-azahexacyclo[7.7.2.1²,⁵.0¹,¹⁰.0³,⁸.0¹³,¹⁷]nonadecan-11-ium bromide
Synonyms: (2S,3S,5S,8S,10R,17R)-13-[2-(2-acetamidophenyl)-2-oxoethyl]-3,5,8-trihydroxy-4,6,16-trimethoxy-11-methyl-11-azahexacyclo[7.7.2.1²,⁵.0¹,¹⁰.0³,⁸.0¹³,¹⁷]nonadecan-11-ium bromide
Molecular Weight (g/mol): 665.62
Molecular Formula: C32H45BrN2O8
InChi Key: DUHCHCGKLJHQCG-JOJFFJMOSA-N
SMILES: [Br-].COC1[C@]2(O)C[C@@H]3[C@@]1(O)[C@](O)(CC2OC)C1C[C@H]2C33[C@@H]1[NH+](C)CC2(CC(=O)C1=CC=CC=C1NC(C)=O)CCC3OC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results