missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ LanthaScreen™ Tb-anti-Histone H3K4me2 Antibody
The LanthaScreen™ Tb-anti-Histone H3K4me2 antibody is a key component of the BacMam Histone H3K4me2 Cellullar Assay (A14164).
$18490.00 - $45110.00
Specifications
| Conjugate | Tb (Terbium)/Lanthanide Chelates |
|---|---|
| Concentration | 3500 nM, 0.5 mg/mL |
| Form | Liquid |
| Shipping Condition | Dry Ice |
| Target Modification State | Methylated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
A14158
|
Thermo Scientific™
A14158 |
100 μg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
A14153
|
Thermo Scientific™
A14153 |
1 mg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- The LanthaScreen™ Tb-anti-Histone H3K4me2 antibody is a key component of the BacMam Histone H3K4me2 Cellullar Assay (A14164)
- When this terbium (Tb)- labeled antibody is paired with baculovirus-mediated (BacMam) expression of a Histone H3-GFP fusion protein, the TR-FRET readout enables sensitive detection of H3 lysine 4 dimethylation
- The resulting high-throughput screening (HTS) compatible assay can be used to identify inhibitors of methyltransferases and demethylases acting on lysine 4 of histone H3 in the cell type of your choice
- For research use only
- Not for human or animal therapeutic or diagnostic use
LanthaScreen™ Tb-anti-Histone H3K4me2 Antibody Allows Users to:
- Identify more relevant inhibitors since the methyl transferases and demethylases are in their natural protein complexes
- Conserve precious cell samples with the miniaturizable homogenous assay format
- Improve data quality with the advantages of TR-FRET
Specifications
| Tb (Terbium)/Lanthanide Chelates | |
| Liquid | |
| Methylated |
| 3500 nM, 0.5 mg/mL | |
| Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.