Learn More
Thermo Scientific™ LanthaScreen™ Tb-anti-Histone H3K4me2 Antibody
The LanthaScreen™ Tb-anti-Histone H3K4me2 antibody is a key component of the BacMam Histone H3K4me2 Cellullar Assay (A14164).
Supplier: Thermo Scientific™ A14153
Description
- The LanthaScreen™ Tb-anti-Histone H3K4me2 antibody is a key component of the BacMam Histone H3K4me2 Cellullar Assay (A14164)
- When this terbium (Tb)- labeled antibody is paired with baculovirus-mediated (BacMam) expression of a Histone H3-GFP fusion protein, the TR-FRET readout enables sensitive detection of H3 lysine 4 dimethylation
- The resulting high-throughput screening (HTS) compatible assay can be used to identify inhibitors of methyltransferases and demethylases acting on lysine 4 of histone H3 in the cell type of your choice
- For research use only
- Not for human or animal therapeutic or diagnostic use
LanthaScreen™ Tb-anti-Histone H3K4me2 Antibody Allows Users to:
- Identify more relevant inhibitors since the methyl transferases and demethylases are in their natural protein complexes
- Conserve precious cell samples with the miniaturizable homogenous assay format
- Improve data quality with the advantages of TR-FRET
Specifications
| 3500 nM, 0.5 mg/mL | |
| 1 mg | |
| Liquid | |
| Methylated |
| Tb (Terbium)/Lanthanide Chelates | |
| Store the LanthaScreen™ Tb-anti-histone H3K4me2 Antibody at -20° C. | |
| Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.