missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) sulfide, 99% (REO)

CAS: 12031-49-1 | La2S3 | 373.99 g/mol

Supplier:  Thermo Scientific Chemicals 02265509

 View more versions of this product

Catalog No. AA2265509


Only null left
Explore available promotions

Description

Description

Lanthanum(III) sulfide is used as a precursor in the production of gallium lanthanum sulfide and strontium lathanum sulfide. It is also used in glass manufacturing. It is doped with other metals and used in optical fibers. It is also used as optical switching, high power-delivery, acousto-optic devices, air-guiding fibers, fiber sensors and mid-IR devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Lanthanum(III) sulfide
Pale Yellow
Powder
99% (REO)
MFCD00016168
YTYSNXOWNOTGMY-UHFFFAOYSA-N
lanthanum(3+);trisulfide
165996
Like rotten eggs
2100° (vacuum)
12031-49-1
10 g
La2S3
lanthanum sulfide, dilanthanum trisulfide, dilanthanum trisulphide, lanthanum sesquisulfide, lanthanum 3+ sulfide, lanthanum sulfide la10s15, lanthanum disulfide, lanthanum 3+ trisulfide
[S--].[S--].[S--].[La+3].[La+3]
373.99
374.02
4.911

Chemical Identifiers

12031-49-1
373.99
YTYSNXOWNOTGMY-UHFFFAOYSA-N
165996
[S--].[S--].[S--].[La+3].[La+3]
La2S3
MFCD00016168
lanthanum sulfide, dilanthanum trisulfide, dilanthanum trisulphide, lanthanum sesquisulfide, lanthanum 3+ sulfide, lanthanum sulfide la10s15, lanthanum disulfide, lanthanum 3+ trisulfide
lanthanum(3+);trisulfide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P231+P232-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H261-H315-H319-H335

DOTInformation : DOT Class: 4.3, Packaging Group: III

EINECSNumber : 234-752-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only