missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) sulfide, 99% (REO)

Catalog No. p-4824774
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA2265504 2 g
AA2265509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA2265504 Supplier Thermo Scientific Chemicals Supplier No. 02265504

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 12031-49-1 | La2S3 | 373.99 g/mol

Lanthanum(III) sulfide is used as a precursor in the production of gallium lanthanum sulfide and strontium lathanum sulfide. It is also used in glass manufacturing. It is doped with other metals and used in optical fibers. It is also used as optical switching, high power-delivery, acousto-optic devices, air-guiding fibers, fiber sensors and mid-IR devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12031-49-1
Molecular Formula La2S3
Molecular Weight (g/mol) 373.99
MDL Number MFCD00016168
InChI Key YTYSNXOWNOTGMY-UHFFFAOYSA-N
Synonym lanthanum sulfide, dilanthanum trisulfide, dilanthanum trisulphide, lanthanum sesquisulfide, lanthanum 3+ sulfide, lanthanum sulfide la10s15, lanthanum disulfide, lanthanum 3+ trisulfide
PubChem CID 165996
IUPAC Name lanthanum(3+);trisulfide
SMILES [S--].[S--].[S--].[La+3].[La+3]

Specifications

Melting Point 2100° (vacuum)
Color Pale Yellow
Physical Form Powder
Quantity 2 g
Odor Like rotten eggs
Assay Percent Range 99% (REO)
Density 4.911
Formula Weight 374.02
Chemical Name or Material Lanthanum(III) sulfide
Hazard Category H261-H315-H319-H335
Precautionary Statement P231+P232-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation DOT Class: 4.3, Packaging Group: III
EINECSNumber 234-752-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only