accessibility menu, dialog, popup

UserName

CAS RN 12031-49-1

IUPAC Name: dilanthanum(3+) trisulfanediide
Synonyms: dilanthanum(3+) trisulfanediide
Molecular Weight (g/mol): 373.99
Molecular Formula: La2S3
InChi Key: YTYSNXOWNOTGMY-UHFFFAOYSA-N
SMILES: [S--].[S--].[S--].[La+3].[La+3]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results