missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) chloride heptahydrate, Puriss. p.a., ≥99%, Solstice

Catalog No. 60046051
Change view
Click to view available options
Quantity:
1 Ea.
6 x 1 Ea.
100 g
6 x 100 g
Packaging:
Ampoule
Poly bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046051 6 x 1 Ea. Ampoule
6002390 100 g Poly bottle
60045978 6 x 100 g Poly bottle
6002403 1 Ea. Ampoule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60046051 Supplier Solstice Supplier No. 320386X1EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

for 1L standard solution

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 10025-84-0 , 7732-18-5
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710

Specifications

CAS Min % ≥90.0000%, ≥5.0000%
Physical Form Concentrate
CAS Max % ≤100.0000%, <10.0000%
Quantity 6 x 1 Ea.
Packaging Ampoule
Density 1.00 g/cm3
Molecular Formula ClNa
Linear Formula NaCl
MDL Number MFCD00003477
UN Number NONH for all modes of transport
Beilstein 3534976
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES [Na+].[Cl-]
IUPAC Name sodium chloride
Molecular Weight (g/mol) 58.44
Formula Weight 58.44
Grade Volumetric
Chemical Name or Material Sodium chloride concentrate, For 1L standard solution
Show More Show Less
ShelfLife 1260 days from date of manufacture
EINECSNumber 231-791-2
Recommended Storage Room Temp