accessibility menu, dialog, popup

UserName

H2OH2OH2OH2OH2OH2OH2OClClClLa

CAS RN 10025-84-0

IUPAC Name: lanthanum(3+) heptahydrate trichloride
Synonyms: lanthanum chloride
Molecular Weight (g/mol): 371.36
Molecular Formula: Cl3H14LaO7
InChi Key: FDFPDGIMPRFRJP-UHFFFAOYSA-K
SMILES: O.O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[La+3]
Molecular Weight (g/mol) 
  • (10)
  • (2)
Quantity 
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
Grade 
  • (2)
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (1)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (7)
  • (10)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)