missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum chloride, hydrated, 64.5-70% LaCl3

CAS: 20211-76-1 | Cl3La | 245.26 g/mol

$176.64 - $2787.44

Chemical Identifiers

CAS 20211-76-1
Molecular Formula Cl3La
Molecular Weight (g/mol) 245.26
MDL Number MFCD00149753
InChI Key ICAKDTKJOYSXGC-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[La+3]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC199151000
SDS View Documents
Thermo Scientific Chemicals
199151000
100 g Glass bottle
Each for $176.64
Only null left
 
AC199155000
SDS View Documents
Thermo Scientific Chemicals
199155000
500 g Glass bottle
Each for $611.03
Only null left
 
AC199150025
SDS View Documents
Thermo Scientific Chemicals
199150025
2.5 kg Glass bottle
Each for $2,787.44
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

20211-76-1
245.26
ICAKDTKJOYSXGC-UHFFFAOYSA-K
[Cl-].[Cl-].[Cl-].[La+3]
Cl3La
MFCD00149753
lanthanum(3+) trichloride

Specifications

White
Adhering Crystals
Cl3La
MFCD00149753
Solubility in water: soluble
[Cl-].[Cl-].[Cl-].[La+3]
245.26
64.5 to 70% LaCl3
Lanthanum chloride, hydrated
20211-76-1
100 g
LaCl3·xH2O
10,5193
ICAKDTKJOYSXGC-UHFFFAOYSA-K
lanthanum(3+) trichloride
245.27
Glass bottle
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.
May be corrosive to metals.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only