missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum chloride, hydrated, 64.5-70% LaCl3

CAS: 20211-76-1 | Cl3La | 245.26 g/mol

Supplier:  Thermo Scientific Chemicals 199155000

 View more versions of this product

Catalog No. AC199155000

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Lanthanum chloride, hydrated
20211-76-1
500 g
LaCl3·xH2O
10,5193
ICAKDTKJOYSXGC-UHFFFAOYSA-K
lanthanum(3+) trichloride
245.27
Glass bottle
White
Adhering Crystals
Cl3La
MFCD00149753
Solubility in water: soluble
[Cl-].[Cl-].[Cl-].[La+3]
245.26
64.5 to 70% LaCl3

Chemical Identifiers

20211-76-1
245.26
ICAKDTKJOYSXGC-UHFFFAOYSA-K
[Cl-].[Cl-].[Cl-].[La+3]
Cl3La
MFCD00149753
lanthanum(3+) trichloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.
May be corrosive to metals.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only