missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Lanosterol, 50%

CAS: 79-63-0 | C30H50O, C30H50O | 426.73 g/mol

$175.78 - $489.86

Chemical Identifiers

CAS 79-63-0
Molecular Formula C30H50O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC466720050
View Documents
Thermo Scientific Chemicals
466720050
5 g
Each for $175.78
Only null left
 
AC466720250
View Documents
Thermo Scientific Chemicals
466720250
25 g
Each for $489.86
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

79-63-0
C30H50O

Specifications

79-63-0
5 g
C30H50O
CAHGCLMLTWQZNJ-BQNIITSRSA-N
(1R,3aR,5aR,7S,9aS,11aR)-3a,6,6,9a,11a-pentamethyl-1-[(2R)-6-methylhept-5-en-2-yl]-1H,2H,3H,3aH,4H,5H,5aH,6H,7H,8H,9H,9aH,10H,11H,11aH-cyclopenta[a]phenanthren-7-ol
Lanosterol
50%
137.0°C
426.73
C[C@H](CCC=C(C)C)[C@H]1CC[C@@]2(C)C3=C(CC[C@]12C)[C@@]1(C)CC[C@H](O)C(C)(C)[C@@H]1CC3
426.73
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 201-214-9

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only