missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LaMotte™ BioPaddles™ Dual-Agar Paddles for Food and Beverage
Contain microbe-specific media enclosed in a sterile vial
$184.50 - $184.50
Specifications
| Content And Storage | 18°C to 25°C |
|---|---|
| Manufacturing Origin | United States |
| Sterility | Sterile |
| Includes | Free basic microbe identification application and access to technical information sheet |
| Packaging Type | Paddle |
Includes: Free basic microbe identification app. and access to technical information sheet.
| Catalog Number | Mfr. No. | Quantity | Media Type | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Media Type | Price | Quantity | ||||
|
03-420-897
|
LaMotte™
5550 |
40 mL | NA/NA dual Agar |
Each for $184.50
|
|
||||
|
03-420-898
|
LaMotte™
5551 |
40 mL | SAB/SAB dual Agar |
Each for $184.50
|
|
||||
Description
Flexible, ready-to-use paddles identify and quantify microbes in air, soil, water or on any surface.
- Longer shelf life than traditional Petri dishes
- No refrigeration required
- No need for inoculating loops or Bunsen burners
- Requires simple incubation and a magnifier
- Unique design enhances colony morphology characterization
- Ten paddles per box
Nutrient-TTC/EMB: For the cultivation and enumeration of Coliform bacteria including TCC (Total Colony Counts). Non-selective, supports growth of a wide variety of organisms.
LIS/EMB Agar: For enumeration and selective isolation of Listeria spp. and total coliform (TCC)
HEK/SS: For enumeration and selective isolation of pathogenic intestinal bacteria.
MacConkey/EMB:For isolation and differentiation of Gram (-) cocci enteric bacilli, coliform, and recovery of stressed coliforms.
Specifications
| 18°C to 25°C | |
| Sterile | |
| Paddle |
| United States | |
| Free basic microbe identification application and access to technical information sheet | |
| 40 mL |