missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LaMotte™ BioPaddles™ Dual-Agar Paddles for Food and Beverage
Description
Includes
Free basic microbe identification app. and access to technical information sheet.
Flexible, ready-to-use paddles identify and quantify microbes in air, soil, water or on any surface.
- Longer shelf life than traditional Petri dishes
- No refrigeration required
- No need for inoculating loops or Bunsen burners
- Requires simple incubation and a magnifier
- Unique design enhances colony morphology characterization
- Ten paddles per box
Nutrient-TTC/EMB: For the cultivation and enumeration of Coliform bacteria including TCC (Total Colony Counts). Non-selective, supports growth of a wide variety of organisms.
LIS/EMB Agar: For enumeration and selective isolation of Listeria spp. and total coliform (TCC)
HEK/SS: For enumeration and selective isolation of pathogenic intestinal bacteria.
MacConkey/EMB:For isolation and differentiation of Gram (-) cocci enteric bacilli, coliform, and recovery of stressed coliforms.
Specifications
Specifications
| Content And Storage | 18°C to 25°C |
| Manufacturing Origin | United States |
| Media Type | NA/NA dual Agar |
| Sterility | Sterile |
| For Use With (Application) | Food and beverage: cultivation of a wide variety of non-fastidious bacteria |
| Includes | Free basic microbe identification application and access to technical information sheet |
| Packaging Type | Paddle |
| Quantity | 40 mL |