missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LaMotte™ BioPaddles™ Dual-Agar Paddles for Food and Beverage
Contain microbe-specific media enclosed in a sterile vial
Supplier: LaMotte™ 5551
Includes: Free basic microbe identification app. and access to technical information sheet.
Description
Flexible, ready-to-use paddles identify and quantify microbes in air, soil, water or on any surface.
- Longer shelf life than traditional Petri dishes
- No refrigeration required
- No need for inoculating loops or Bunsen burners
- Requires simple incubation and a magnifier
- Unique design enhances colony morphology characterization
- Ten paddles per box
Nutrient-TTC/EMB: For the cultivation and enumeration of Coliform bacteria including TCC (Total Colony Counts). Non-selective, supports growth of a wide variety of organisms.
LIS/EMB Agar: For enumeration and selective isolation of Listeria spp. and total coliform (TCC)
HEK/SS: For enumeration and selective isolation of pathogenic intestinal bacteria.
MacConkey/EMB:For isolation and differentiation of Gram (-) cocci enteric bacilli, coliform, and recovery of stressed coliforms.
Specifications
| 18°C to 25°C | |
| SAB/SAB dual Agar | |
| Food and beverage: selective isolation of Fungi (Yeasts and molds) (SAB) | |
| Paddle |
| United States | |
| Sterile | |
| Free basic microbe identification application and access to technical information sheet | |
| 40 mL |