missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Lambda Exonuclease (10 U/μL)

Catalog No. FEREN0562
Change view
Click to view available options
Quantity:
1,000 units
5,000 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEREN0562 5,000 units
FEREN0561 1,000 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEREN0562 Supplier Thermo Scientific™ Supplier No. EN0562

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Lambda Exonuclease is a highly processive exodeoxyribonuclease that selectively digests the 5'-phosphorylated strand of double-stranded DNA in 5'-3' direction.

Thermo Scientific Lambda Exonuclease is a highly processive 5'≥3' exodeoxyribonuclease. It selectively digests the 5'-phosphorylated strand of double-stranded DNA. The enzyme exhibits low activity on single-stranded DNA and non-phosphorylated DNA, and has no activity at nicks and limited activity at gaps in DNA.

Highlights

  • Active in Thermo Scientific PCR buffers

Applications

  • Generating single-stranded PCR products for use in:
    • DNA sequencing
    • Analysis of DNA single-strand conformation polymorphism (SSCP)
    • Rolling circle amplification
     
  • Producing single-stranded DNA from double-stranded DNA fragments
  • Cloning of PCR products

Note

Use of this enzyme in certain applications may be covered by patents and may require a license.

Specifications

Concentration 10 U/μL
Enzyme Exonuclease
Quantity 5,000 units
Product Type Exonuclease

For Research Use Only. Not for use in diagnostic procedures.