missing translation for 'onlineSavingsMsg'
Learn More

Lactitol monohydrate, 98%

CAS: 81025-04-9 | C12H26O12 | 362.328 g/mol

$113.16 - $402.16

Chemical Identifiers

CAS 81025-04-9
Molecular Formula C12H26O12
Molecular Weight (g/mol) 362.328
MDL Number MFCD00150767
InChI Key LXMBXZRLTPSWCR-XBLONOLSSA-N
Synonym lactitol monohydrate, d-lactitol monohydrate, lactitol hydrate, unii-uh2k6w1y64, 4-o-beta-d-galactopyranosyl-d-glucitol monohydrate, d-glucitol, 4-o-beta-d-galactopyranosyl-, monohydrate, 4-o-beta-d-galactopyranosyl-d-glucitol, portolac tn, 2s,3r,4r,5r-4-2s,3r,4s,5r,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxyhexane-1,2,3,5,6-pentol;hydrate
PubChem CID 3067270
IUPAC Name (2S,3R,4R,5R)-4-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhexane-1,2,3,5,6-pentol;hydrate
SMILES C(C1C(C(C(C(O1)OC(C(CO)O)C(C(CO)O)O)O)O)O)O.O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAB2101603
View Documents Promotion Details
N/A 1 g
Each for $113.16
Only null left
 
AAB2101606
View Documents Promotion Details
N/A 5 g
Each for $402.16
Only null left
 
Description

Description

Lactitol monohydrate is used in low fat foods, anthelmintic, cosmetic and pharmaceutical excipients. It is used in sugar-free candies, cookies, chocolate and ice cream. It is a laxative, which used to treat constipation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lactitol monohydrate is used in low fat foods, anthelmintic, cosmetic and pharmaceutical excipients. It is used in sugar-free candies, cookies, chocolate and ice cream. It is a laxative, which used to treat constipation.

Solubility
Soluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

81025-04-9
362.328
LXMBXZRLTPSWCR-XBLONOLSSA-N
3067270
C(C1C(C(C(C(O1)OC(C(CO)O)C(C(CO)O)O)O)O)O)O.O
C12H26O12
MFCD00150767
lactitol monohydrate, d-lactitol monohydrate, lactitol hydrate, unii-uh2k6w1y64, 4-o-beta-d-galactopyranosyl-d-glucitol monohydrate, d-glucitol, 4-o-beta-d-galactopyranosyl-, monohydrate, 4-o-beta-d-galactopyranosyl-d-glucitol, portolac tn, 2s,3r,4r,5r-4-2s,3r,4s,5r,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxyhexane-1,2,3,5,6-pentol;hydrate
(2S,3R,4R,5R)-4-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhexane-1,2,3,5,6-pentol;hydrate

Specifications

81025-04-9
C12H26O12
14,5340
Soluble in water.
C(C1C(C(C(C(O1)OC(C(CO)O)C(C(CO)O)O)O)O)O)O.O
362.328
362.33 (344.31 Anhydrous)
Lactitol monohydrate
95°C to 98°C
MFCD00150767
lactitol monohydrate, d-lactitol monohydrate, lactitol hydrate, unii-uh2k6w1y64, 4-o-beta-d-galactopyranosyl-d-glucitol monohydrate, d-glucitol, 4-o-beta-d-galactopyranosyl-, monohydrate, 4-o-beta-d-galactopyranosyl-d-glucitol, portolac tn, 2s,3r,4r,5r-4-2s,3r,4s,5r,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxyhexane-1,2,3,5,6-pentol;hydrate
LXMBXZRLTPSWCR-XBLONOLSSA-N
(2S,3R,4R,5R)-4-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhexane-1,2,3,5,6-pentol;hydrate
3067270
98%
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only