missing translation for 'onlineSavingsMsg'
Learn More

Lactic Acid, 0.3% (w/v), Ricca Chemical

Catalog No. R4263000100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R4263000100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R4263000100 Supplier Ricca Chemical Company Supplier No. R4263000100A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 50-21-5 , 7732-18-5
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
MDL Number MFCD00004520
InChI Key JVTAAEKCZFNVCJ-UHFFFAOYNA-N
PubChem CID 612
ChEBI CHEBI:78320
IUPAC Name 2-hydroxypropanoic acid
SMILES CC(O)C(O)=O

Specifications

Packaging Natural Poly Bottle
CAS Min % 98.0, 0.3
CAS Max % 100.0, 0.31
Physical Form Liquid
Color Colorless
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Standard Solution
Type Standard
Quantity 100 mL
Percent Purity 0.003
Grade Laboratory
Chemical Name or Material Lactic Acid Standard, 3% w/v
Show More Show Less