Learn More
MilliporeSigma™ Labscale TFF System Reservoir
Catalog No. XX42RES01
Change view
Click to view available options
Type:
Reservoir
Catalog No. Type
XX42RES01 Reservoir
1 options
Catalog No. XX42RES01 Supplier MilliporeSigma™ Supplier No. XX42RES01
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
Graduated 500mL acrylic reservoir with retentate back pressure valve, feed and retentate pressure indicators; stirrer assembly; diaphragm pump assembly with tubing and power supply.
Easy to use for sample preparation
Complete linear, scalable solution for small-volume processing designed specifically to use the Pellicon™ XL Device for simple sample prep and more complex scale-up work.
  • 500mL reservoir accepts direct docking of the Pellicon XL device, eliminating need for tubing connections
Order Info
Individual components are also available for a custom small-volume TFF system. Pellicon™ XL devices sold separately.
Specifications
Material Acrylic
For Use With (Application) Accepts direct docking of the Pellicon™ XL device, eliminating need for tubing connections
Includes Retentate back pressure valve
Quantity 1/Pk.
Type Reservoir
Volume (Metric) 500 mL
For Use With (Equipment) Labscale TFF System