Learn More
Labconco™ Sample Blocks for RapidVap™ N2/48 Evaporation Systems
$6080.48 - $6261.29
Products 2
Catalog Number Mfr. No. Holds Price Quantity  
Catalog Number Mfr. No. Holds Price Quantity  
16-317-68
View Documents
Labconco™
7482400
35 mL Tubes
Each for $6,261.29
Only null left
16-317-66
View Documents
Labconco™
7482200
10 mL Tubes
Each for $6,080.48
Only null left
Description
Description
  • Four different interchangeable aluminum accessory blocks, coated with PTFE
  • Sample blocks hold either 48 smaller (4.5 mL to 26 mL) or eight larger (450 mL) samples
  • Glassware for all blocks sold separately
†For tubes up to maximum capacity shown. Actual sample volume is 66–75% total tube capacity and depends on tube shape, height and vortex speed.
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'