missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Diaphragm Pumps For Precise™ Basic™ and Controlled Atmosphere Glove Boxes

For drying train circulation

$4,169.81 - $4,479.30

Specifications

For Use With (Equipment) Protector Combination Glove Box
Weight (English) 11 lb.
Weight (Metric) 5 Kg
Material Die-cast Aluminum
For Use With (Application) Drying Train Circulation
View More Specs

Products 2
missing translation for 'productCode' missing translation for 'brand' Electrical Requirements Price Quantity & Availability  
missing translation for 'productCode' missing translation for 'brand' Electrical Requirements Price Quantity & Availability  
16108379
View Documents Promotion Details
Labconco™
5248201
230 V, 50 Hz
Each for $4,479.30
Only null left
 
16108378
View Documents Promotion Details
Labconco™
5248200
115 V, 60 Hz
Each for $4,169.81
Only null left
 
Description

Description

  • Consists of single cylinder, oil-less diaphragm pumps with permanently lubricated bearings
  • Mounted on rubber feet
  • Provides 206mBar ultimate vacuum
  • 1/8 hp motor, Die cast aluminum
  • 1/4 in. diameter inlet and outlet connections

Specifications

Specifications

Protector Combination Glove Box
5 Kg
Drying Train Circulation
11 lb.
Die-cast Aluminum
Videos
SDS
Documents

Documents