missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ Phosphate Buffered Saline, powder for a 1L 10X stock solution
Description
- For in vitro diagnostic use
- 1X working solution contains 0.14M Sodium chloride, 0.003M Potassium chloride, 0.002M Potassium phosphate, 0.01M Sodium phosphate
- Store at room temperature.
- Stable for up to three years
Specifications
Specifications
| Chemical Name or Material | Phosphate Buffered Saline (PBS) |
| Concentration | 10 X |
| For Use With (Application) | Vitro Diagnostic |
| Quantity | 1 L |
| Solution Type | Phosphate Buffered Saline |
For Research Use only