missing translation for 'onlineSavingsMsg'
Learn More

Eagle™ Lab Pack Open Head Poly Drum with Bung Holes, 14 gal.
GREENER_CHOICE

Drum is constructed of blow-molded high-density polyethylene (HDPE) with UV inhibitors, is durable and lightweight, as well as weather and chemical resistant. Bung lid includes two holes (3/4 in. and 2 in. diameters) and is ideal for safe storage, as well as handling or transportation of hazardous materials.

$99.11 - $105.86

Specifications

Product Type Drum
Closure Included Yes
Capacity (English) 14 gal.
Capacity (Metric) 53.06 L
For Use With (Application) Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration
View More Specs

Products 6
Catalog Number Mfr. No. Color Closure Type Price Quantity & Availability  
Catalog Number Mfr. No. Color Closure Type Price Quantity & Availability  
19338433
GREENER_CHOICE
View Documents Promotion Details
Eagle™
1610MBKBRBG1
Black Metal Bolt Ring
Each for $102.88
Only null left
 
19338432
GREENER_CHOICE
View Documents Promotion Details
Eagle™
1610MBBRBG1
Blue Metal Bolt Ring
Each for $105.86
Only null left
 
19338431
GREENER_CHOICE
View Documents Promotion Details
Eagle™
1610MBRBG1
Yellow Metal Bolt Ring
Each for $105.86
Only null left
 
19338430
GREENER_CHOICE
View Documents Promotion Details
Eagle™
1610MBKBG1
Black Metal Lever-lock
Each for $99.11
Only null left
 
19338429
GREENER_CHOICE
View Documents Promotion Details
Eagle™
1610MBBG1
Blue Metal Lever-lock
Each for $101.95
Only null left
 
19338428
GREENER_CHOICE
View Documents Promotion Details
Eagle™
1610MBG1
Yellow Metal Lever-lock
Each for $101.95
Only null left
 
Description

Description

  • Constructed from blow-molded high-density polyethylene (HDPE) with UV inhibitors
  • For use with smaller non-damaged bottles, cans, carboys and 5 gal. pails of non-leaking hazardous materials
  • Bung lid includes two holes, one of each diameter: 0.75 in. and 2 in.
  • If used with leaking materials, drums must be packed with sufficient sorbents
  • No free liquids
Specifications

Specifications

Drum
14 gal.
Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration
Open
63.5 cm
10 lb.
Yes
53.06 L
HDPE
25 in.
0.75 in., 2 in. (Bung Holes)
Videos
SDS
Documents

Documents