missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eagle™ Lab Pack Open Head Poly Drum with Bung Holes, 14 gal.

Drum is constructed of blow-molded high-density polyethylene (HDPE) with UV inhibitors, is durable and lightweight, as well as weather and chemical resistant. Bung lid includes two holes (3/4 in. and 2 in. diameters) and is ideal for safe storage, as well as handling or transportation of hazardous materials.
Supplier: Eagle™ 1610MBG1
Description
- Constructed from blow-molded high-density polyethylene (HDPE) with UV inhibitors
- For use with smaller non-damaged bottles, cans, carboys and 5 gal. pails of non-leaking hazardous materials
- Bung lid includes two holes, one of each diameter: 0.75 in. and 2 in.
- If used with leaking materials, drums must be packed with sufficient sorbents
- No free liquids
Chemical Identifiers
| Drum | |
| 14 gal. | |
| Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration | |
| Open | |
| 63.5 cm | |
| 10 lb. |
| Yes | |
| 53.06 L | |
| HDPE | |
| 25 in. | |
| 0.75 in., 2 in. (Bung Holes) |
Specifications
| Drum | |
| Metal Lever-lock | |
| 53.06 L | |
| Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration | |
| Open | |
| 63.5 cm | |
| 10 lb. |
| Yes | |
| 14 gal. | |
| Yellow | |
| HDPE | |
| 25 in. | |
| 0.75 in., 2 in. (Bung Holes) |