missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-(-)-Xylose 97.0+%, TCI America™
Supplier: TCI America X002125G
| Quantity | 25 g |
|---|
Chemical Identifiers
| C5H10O5 | |
| MFCD00151096 | |
| l-xylose, l +-xylose, l---xylose, 2s,3r,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-xylose, l-xylo-pentose, l-xyl, xylose, l, linear l-xylose, l--xylose | |
| CHEBI:17979 | |
| OCC(O)C(O)C(O)C=O |
Specifications
| L-(-)-Xylose | |
| 150°C | |
| C5H10O5 | |
| MFCD00151096 | |
| PYMYPHUHKUWMLA-UHFFFAOYNA-N | |
| 2,3,4,5-tetrahydroxypentanal | |
| 95259 | |
| 150.13 | |
| Crystalline Powder |
| 609-06-3 | |
| White | |
| 25 g | |
| l-xylose, l +-xylose, l---xylose, 2s,3r,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-xylose, l-xylo-pentose, l-xyl, xylose, l, linear l-xylose, l--xylose | |
| OCC(O)C(O)C(O)C=O | |
| 150.13 | |
| CHEBI:17979 | |
| ≥97.0% (T) |
Safety and Handling
EINECSNumber : (9)-0318
TSCA : Yes