missing translation for 'onlineSavingsMsg'
Learn More

L-(-)-Xylose 97.0+%, TCI America™

Catalog No. p-7137958
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X00215G 5 g
X002125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. X00215G Supplier TCI America Supplier No. X00215G

Chemical Identifiers

CAS 609-06-3
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
MDL Number MFCD00151096
InChI Key PYMYPHUHKUWMLA-UHFFFAOYNA-N
Synonym l-xylose, l +-xylose, l---xylose, 2s,3r,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-xylose, l-xylo-pentose, l-xyl, xylose, l, linear l-xylose, l--xylose
PubChem CID 95259
ChEBI CHEBI:17979
IUPAC Name 2,3,4,5-tetrahydroxypentanal
SMILES OCC(O)C(O)C(O)C=O

Specifications

Melting Point 150°C
Color White
Quantity 5 g
Formula Weight 150.13
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-(-)-Xylose
EINECSNumber (9)-0318
TSCA Yes