missing translation for 'onlineSavingsMsg'
Learn More

L-Valine, USP, JP, bioCERTIFIED, 98.5-101%, Spectrum™ Chemical

Spectrum™ Chemical bioCERTIFIED products are suitable for biopharmaceutical production.

Chemical Identifiers

CAS 72-18-4
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18614864
View Documents Promotion Details
N/A 1 kg Plastic Bottle N/A
Only null left
 
18614865
View Documents Promotion Details
N/A 12 kg Plastic Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

  • Full testing including bioburden, endotoxin and elemental analysis
  • Comprehensive regulatory and scientific documentation
  • Change control and batch traceabilitiy
  • Biopharm industry standard CFR 210/211 cGMP
Specifications

Chemical Identifiers

72-18-4

Specifications

0.02%
2 ppm
0.021%
To pass test
0.2%
0.1%
98.5 to 101%
L-Valine
72-18-4
6.0 EU/g
20 ppm
10 ppm
To pass test
0.03%
USP/JP/bioCERTIFIED
Videos
SDS
Documents

Documents