missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-(-)-Tyrosine 98.5+%, TCI America™
$96.84 - $1,063.23
Chemical Identifiers
| CAS | 60-18-4 |
|---|---|
| Molecular Formula | C9H11NO3 |
| Molecular Weight (g/mol) | 181.19 |
| MDL Number | MFCD00002606 |
| InChI Key | OUYCCCASQSFEME-SVGMAFHSNA-N |
| Synonym | l-tyrosine, tyrosine, s-tyrosine, p-tyrosine, h-tyr-oh, l-p-tyrosine, 2s-2-amino-3-4-hydroxyphenyl propanoic acid, 4-hydroxy-l-phenylalanine, l---tyrosine, tyrosine, l |
| PubChem CID | 6057 |
| ChEBI | CHEBI:17895 |
| IUPAC Name | (2S)-2-amino-3-(4-hydroxyphenyl)propanoic acid |
| SMILES | N[C@@H](CC1=CC=C(O)C=C1)C(O)=O |
Chemical Identifiers
| C9H11NO3 | |
| MFCD00002606 | |
| l-tyrosine, tyrosine, s-tyrosine, p-tyrosine, h-tyr-oh, l-p-tyrosine, 2s-2-amino-3-4-hydroxyphenyl propanoic acid, 4-hydroxy-l-phenylalanine, l---tyrosine, tyrosine, l | |
| CHEBI:17895 | |
| N[C@@H](CC1=CC=C(O)C=C1)C(O)=O |
Specifications
| 60-18-4 | |
| C9H11NO3 | |
| l-tyrosine, tyrosine, s-tyrosine, p-tyrosine, h-tyr-oh, l-p-tyrosine, 2s-2-amino-3-4-hydroxyphenyl propanoic acid, 4-hydroxy-l-phenylalanine, l---tyrosine, tyrosine, l | |
| N[C@@H](CC1=CC=C(O)C=C1)C(O)=O | |
| 181.19 | |
| CHEBI:17895 | |
| ≥98.5% (T) | |
| L-(-)-Tyrosine |
| White | |
| MFCD00002606 | |
| OUYCCCASQSFEME-SVGMAFHSNA-N | |
| (2S)-2-amino-3-(4-hydroxyphenyl)propanoic acid | |
| 6057 | |
| 181.19 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (9)-1596
RTECSNumber : YP2275600
TSCA : Yes