missing translation for 'onlineSavingsMsg'
Learn More

L-(-)-Tyrosine 98.5+%, TCI America™

Catalog No. T055025G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T055025G 25 g
T0550100G 100 g
T0550500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T055025G Supplier TCI America Supplier No. T055025G

Chemical Identifiers

CAS 60-18-4
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.19
MDL Number MFCD00002606
InChI Key OUYCCCASQSFEME-SVGMAFHSNA-N
Synonym l-tyrosine, tyrosine, s-tyrosine, p-tyrosine, h-tyr-oh, l-p-tyrosine, 2s-2-amino-3-4-hydroxyphenyl propanoic acid, 4-hydroxy-l-phenylalanine, l---tyrosine, tyrosine, l
PubChem CID 6057
ChEBI CHEBI:17895
IUPAC Name (2S)-2-amino-3-(4-hydroxyphenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(O)C=C1)C(O)=O

Specifications

Color White
Quantity 25 g
Formula Weight 181.19
Percent Purity ≥98.5% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-(-)-Tyrosine
EINECSNumber (9)-1596
RTECSNumber YP2275600
TSCA Yes