missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Threonine, USP, JP, bioCERTIFIED, 98-101.5%, Spectrum™ Chemical
Spectrum™ Chemical bioCERTIFIED products are suitable for biopharmaceutical production.
$6,573.38 - $58,298.34
Chemical Identifiers
| CAS | 72-19-5 |
|---|---|
| Molecular Formula | C4H9NO3 |
| Molecular Weight (g/mol) | 119.12 |
| InChI Key | AYFVYJQAPQTCCC-UHFFFAOYNA-N |
| IUPAC Name | 2-amino-3-hydroxybutanoic acid |
| SMILES | CC(O)C(N)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
18614862
|
N/A | 1 kg | Plastic Bottle |
Each for $6,573.38
|
|
||||
|
18614863
|
N/A | 12 kg | Plastic Pail |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- Full testing including endotoxin and elemental analysis
- Comprehensive regulatory and scientific documentation
- Change control and batch traceabilitiy
- Biopharm industry standard CFR 210/211 cGMP
Chemical Identifiers
| 72-19-5 | |
| 119.12 | |
| 2-amino-3-hydroxybutanoic acid |
| C4H9NO3 | |
| AYFVYJQAPQTCCC-UHFFFAOYNA-N | |
| CC(O)C(N)C(O)=O |
Specifications
| 72-19-5 | |
| 2 ppm | |
| Clear/Colorless | |
| 0.021% | |
| To pass test | |
| 0.2% | |
| To pass test | |
| -26.0° to -29.0° | |
| AYFVYJQAPQTCCC-UHFFFAOYNA-N | |
| 2-amino-3-hydroxybutanoic acid | |
| 98 to 101.5% |
| 0.02% | |
| 5.2 to 6.2 | |
| 6.0 EU/g | |
| 20 ppm | |
| 10 ppm | |
| C4H9NO3 | |
| 0.1% | |
| 0.03% | |
| CC(O)C(N)C(O)=O | |
| 119.12 | |
| USP/JP/bioCERTIFIED |