missing translation for 'onlineSavingsMsg'
Learn More

L-Threonine, USP, JP, bioCERTIFIED, 98-101.5%, Spectrum™ Chemical

Catalog No. 18614862
Change view
Click to view available options
Quantity:
1 kg
12 kg
Packaging:
Plastic Bottle
Plastic Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614862 1 kg Plastic Bottle
18614863 12 kg Plastic Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18614862 Supplier Spectrum Chemical Mfg Cor Supplier No. T94001KG
Get Up to $500 in Chemicals with Purchase
View all promotions

Spectrum™ Chemical bioCERTIFIED products are suitable for biopharmaceutical production.

  • Full testing including endotoxin and elemental analysis
  • Comprehensive regulatory and scientific documentation
  • Change control and batch traceabilitiy
  • Biopharm industry standard CFR 210/211 cGMP

Chemical Identifiers

CAS 72-19-5
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
InChI Key AYFVYJQAPQTCCC-UHFFFAOYNA-N
IUPAC Name 2-amino-3-hydroxybutanoic acid
SMILES CC(O)C(N)C(O)=O

Specifications

Ammonium 0.02%
Arsenic (As) 2 ppm
pH 5.2 to 6.2
Color Clear/Colorless
Bacterial Endotoxins 6.0 EU/g
Chlorine (Cl) 0.021%
Heavy Metals (as Pb) 20 ppm
Identification To pass test
Iron (Fe) 10 ppm
Loss on Drying 0.2%
Packaging Plastic Bottle
Residual Solvents To pass test
Residue after Ignition 0.1%
Specific Rotation -26.0° to -29.0°
Sulfate 0.03%
Quantity 1 kg
Percent Purity 98 to 101.5%
Grade USP/JP/bioCERTIFIED
Show More Show Less