missing translation for 'onlineSavingsMsg'
Learn More

L-tert-Leucine 98.0+%, TCI America™

Supplier:  TCI America L014725G

 View more versions of this product

Catalog No. L014725G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

L-tert-Leucine
White
MFCD00064218
l-tert-leucine, 2s-2-amino-3,3-dimethylbutanoic acid, 3-methyl-l-valine, l-valine,3-methyl, h-tle-oh, s-2-amino-3,3-dimethylbutanoic acid, 3-methyl-l-valin, l-2-amino-3,3-dimethylbutanoic acid, s-2-amino-3,3-dimethylbutyric acid
CC(C)(C)[C@H](N)C(O)=O
131.18
131.18
Crystalline Powder
20859-02-3
C6H13NO2
25 g
NPDBDJFLKKQMCM-SGAVLPGINA-N
(2S)-2-amino-3,3-dimethylbutanoic acid
164608
≥98.0% (T)

Chemical Identifiers

20859-02-3
131.18
NPDBDJFLKKQMCM-SGAVLPGINA-N
164608
CC(C)(C)[C@H](N)C(O)=O
C6H13NO2
MFCD00064218
l-tert-leucine, 2s-2-amino-3,3-dimethylbutanoic acid, 3-methyl-l-valine, l-valine,3-methyl, h-tle-oh, s-2-amino-3,3-dimethylbutanoic acid, 3-methyl-l-valin, l-2-amino-3,3-dimethylbutanoic acid, s-2-amino-3,3-dimethylbutyric acid
(2S)-2-amino-3,3-dimethylbutanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1632

TSCA : No

SDS
Documents

Documents

Promotions

Promotions