accessibility menu, dialog, popup

UserName

H3C(S)NH2OHOCH3CH3

CAS RN 20859-02-3

IUPAC Name: (2S)-2-amino-3,3-dimethylbutanoic acid
Synonyms: L-tert-leucine
Molecular Weight (g/mol): 131.18
Molecular Formula: C6H13NO2
InChi Key: NPDBDJFLKKQMCM-SGAVLPGINA-N
SMILES: CC(C)(C)[C@H](N)C(O)=O
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (2)
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (3)