missing translation for 'onlineSavingsMsg'
Learn More

L-tert-Leucine 98.0+%, TCI America™

Catalog No. p-7131840
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L01471G 1 g
L01475G 5 g
L014725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. L01471G Supplier TCI America Supplier No. L01471G
Only null left

Chemical Identifiers

CAS 20859-02-3
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00064218
InChI Key NPDBDJFLKKQMCM-SGAVLPGINA-N
Synonym l-tert-leucine, 2s-2-amino-3,3-dimethylbutanoic acid, 3-methyl-l-valine, l-valine,3-methyl, h-tle-oh, s-2-amino-3,3-dimethylbutanoic acid, 3-methyl-l-valin, l-2-amino-3,3-dimethylbutanoic acid, s-2-amino-3,3-dimethylbutyric acid
PubChem CID 164608
IUPAC Name (2S)-2-amino-3,3-dimethylbutanoic acid
SMILES CC(C)(C)[C@H](N)C(O)=O

Specifications

Color White
Quantity 1 g
Formula Weight 131.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-tert-Leucine
EINECSNumber (9)-1632
TSCA No