missing translation for 'onlineSavingsMsg'
Learn More

L-Serine ethyl ester hydrochloride, 99%

CAS: 26348-61-8 | C5H12ClNO3 | 169.61 g/mol

Supplier:  Thermo Scientific Chemicals A1417414

 View more versions of this product

Catalog No. AAA1417414


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Serine ethyl ester hydrochloride is widely used in pharmaceutical and food industry.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents and moisture. Store under dry inert gas. Protect from humidity and water.
Specifications

Specifications

130°C to 132°C
26348-61-8
MFCD00012594
l-serine ethyl ester hydrochloride, h-ser-oet.hcl, ethyl l-serinate hydrochloride, s-ethyl 2-amino-3-hydroxypropanoate hydrochloride, serine ethyl ester hydrochloride, l-serine ethyl ester hcl, l-serine, ethyl ester, hydrochloride, ethyl serinate hydrochloride, ethyl 2s-2-amino-3-hydroxypropanoate hydrochloride, h-ser-oet hcl
JZJQCLZQSHLSFB-UHFFFAOYNA-N
ethyl (2S)-2-amino-3-hydroxypropanoate;hydrochloride
169.61
−4.4° (c=4 in Water)
Hygroscopic
L-Serine ethyl ester hydrochloride
C5H12ClNO3
3562346
Soluble in water.
[H+].[Cl-].CCOC(=O)C(N)CO
25 g
2724185
169.61
99%

Chemical Identifiers

26348-61-8
169.61
JZJQCLZQSHLSFB-UHFFFAOYNA-N
2724185
C5H12ClNO3
MFCD00012594
l-serine ethyl ester hydrochloride, h-ser-oet.hcl, ethyl l-serinate hydrochloride, s-ethyl 2-amino-3-hydroxypropanoate hydrochloride, serine ethyl ester hydrochloride, l-serine ethyl ester hcl, l-serine, ethyl ester, hydrochloride, ethyl serinate hydrochloride, ethyl 2s-2-amino-3-hydroxypropanoate hydrochloride, h-ser-oet hcl
[H+].[Cl-].CCOC(=O)C(N)CO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 247-624-1

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only