accessibility menu, dialog, popup

UserName

HClH3COONH2OH

CAS RN 26348-61-8

IUPAC Name: hydrogen ethyl 2-amino-3-hydroxypropanoate chloride
Synonyms: hydrogen ethyl serinate chloride
Molecular Weight (g/mol): 169.61
Molecular Formula: C5H12ClNO3
InChi Key: JZJQCLZQSHLSFB-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CCOC(=O)C(N)CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)