missing translation for 'onlineSavingsMsg'
Learn More

L-Serine ethyl ester hydrochloride, 99%

Catalog No. p-7021721
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1417406 5 g
AAA1417414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1417406 Supplier Thermo Scientific Chemicals Supplier No. A1417406

missing translation for 'validMainframeMsg'

CAS: 26348-61-8 | C5H12ClNO3 | 169.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Serine ethyl ester hydrochloride is widely used in pharmaceutical and food industry.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents and moisture. Store under dry inert gas. Protect from humidity and water.

Chemical Identifiers

CAS 26348-61-8
Molecular Formula C5H12ClNO3
Molecular Weight (g/mol) 169.61
MDL Number MFCD00012594
InChI Key JZJQCLZQSHLSFB-UHFFFAOYNA-N
Synonym l-serine ethyl ester hydrochloride, h-ser-oet.hcl, ethyl l-serinate hydrochloride, s-ethyl 2-amino-3-hydroxypropanoate hydrochloride, serine ethyl ester hydrochloride, l-serine ethyl ester hcl, l-serine, ethyl ester, hydrochloride, ethyl serinate hydrochloride, ethyl 2s-2-amino-3-hydroxypropanoate hydrochloride, h-ser-oet hcl
PubChem CID 2724185
SMILES [H+].[Cl-].CCOC(=O)C(N)CO

Specifications

Melting Point 130°C to 132°C
Beilstein 3562346
Solubility Information Soluble in water.
IUPAC Name ethyl (2S)-2-amino-3-hydroxypropanoate;hydrochloride
Quantity 5 g
Formula Weight 169.61
Optical Rotation −4.4° (c=4 in Water)
Sensitivity Hygroscopic
Percent Purity 99%
Chemical Name or Material L-Serine ethyl ester hydrochloride
EINECSNumber 247-624-1
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only