missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Serine, Cell Culture Reagent

CAS: 56-45-1 | C3H7NO3 | 105.093 g/mol

Supplier:  Thermo Scientific Chemicals J6218722

 View more versions of this product

Catalog No. AAJ6218722


Only null left

Description

Description

It is a precursor of glycine by serine hydroxymethyltransferase. D-serine, synthesized by serine racemase from L-serine, serves as a neuronal signaling molecule by activating NMDA receptors in the brain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a precursor of glycine by serine hydroxymethyltransferase. D-serine, synthesized by serine racemase from L-serine, serves as a neuronal signaling molecule by activating NMDA receptors in the brain.

Solubility
Soluble in water.

Notes
Store in cool, dry place. Incompatible with oxidizing agents.
Specifications

Specifications

222°C
56-45-1
C3H7NO3
1721404
l-serine, serine, s-2-amino-3-hydroxypropanoic acid, s-serine, beta-hydroxyalanine, h-ser-oh, l-ser, l---serine, serine, l, 2s-2-amino-3-hydroxypropanoic acid
MTCFGRXMJLQNBG-REOHCLBHSA-N
(2S)-2-amino-3-hydroxypropanoic acid
105.093
CHEBI:17115
Cell Culture Reagent
L-Serine
White
MFCD00064224
14,8460
Soluble in water.
C(C(C(=O)O)N)O
100 g
5951
105.09
Powder

Chemical Identifiers

56-45-1
105.093
MTCFGRXMJLQNBG-REOHCLBHSA-N
5951
(2S)-2-amino-3-hydroxypropanoic acid
C3H7NO3
MFCD00064224
l-serine, serine, s-2-amino-3-hydroxypropanoic acid, s-serine, beta-hydroxyalanine, h-ser-oh, l-ser, l---serine, serine, l, 2s-2-amino-3-hydroxypropanoic acid
CHEBI:17115
C(C(C(=O)O)N)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-274-3

RTECSNumber : VT8100000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only